Novus Biologicals
Manufacturer Code:NBP297791AF647
Catalog # NB047785
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human RPGR fragment. |
Conjugate | Alexa Fluor 647 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: COD1 cone dystrophy 1 (X-linked) CORDX1RP3retinitis pigmentosa 15 CRD orf15 PCDX retinitis pigmentosa 3 GTPase regulator retinitis pigmentosa GTPase regulator RP15 X-linked retinitis pigmentosa GTPase regulator XLRP3; retinitis pigmentosa 15; retinitis pigmentosa 3 GTPase regulator; X-linked retinitis pigmentosa GTPase regulator
Gene Aliases: COD1; CORDX1; CRD; orf15; PCDX; RP15; RP3; RPGR; XLRP3
UniProt ID: (Human) Q92834
Entrez Gene ID: (Human) 6103
Molecular Function: DNA binding protein G-protein modulator chromatin/chromatin-binding protein enzyme modulator guanyl-nucleotide exchange factor nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.