Abnova
Manufacturer Code:PAB20072
Catalog # 89-121-573
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human RPGR. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant RPGR.
For Research Use Only
Protein Aliases: COD1; CORDX1; CRD; orf15; PCDX; retinitis pigmentosa 15; retinitis pigmentosa 3 GTPase regulator; RP15; RP3; X-linked retinitis pigmentosa GTPase regulator; XLRP3
Gene Aliases: COD1; CORDX1; CRD; orf15; PCDX; RP15; RP3; RPGR; XLRP3
UniProt ID: (Human) Q92834
Entrez Gene ID: (Human) 6103
Molecular Function: DNA binding protein G-protein modulator chromatin/chromatin-binding protein enzyme modulator guanyl-nucleotide exchange factor nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.