Invitrogen
Manufacturer Code:PA524517
Catalog # PIPA524517
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 187-215 amino acids from the C-terminal region of human RPEL1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hCG2024410; hCG2024410 rcRPE protein hCG&lowbar2024410; Phosphatase and actin regulator 1; rcRPE protein; ribulose-5-phosphate-3-epimerase-like 1; Ribulose-5-phosphate-3-epimerase-like protein 1; Ribulose-phosphate 3-epimerase-like protein 1; RPEL repeat containing 1
Gene Aliases: 9630030F18Rik; dJ257A7.2; KIAA1733; PHACTR1; RPEL; RPEL1
UniProt ID: (Human) Q9C0D0, (Mouse) Q2M3X8, (Human) Q2QD12
Entrez Gene ID: (Human) 221692, (Mouse) 218194, (Human) 729020
Molecular Function: enzyme modulator phosphatase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.