Novus Biologicals
Manufacturer Code:NB100355G
Catalog # NB100355G
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 401.8B11.3D9 |
Immunogen | Bovine RPE65 microsomal membrane proteins. [UniProt# Q28175] |
Conjugate | DyLight 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: All-trans-retinyl-palmitate hydrolase; All-trans-retinyl-palmitate hydrolase EC 3.1.1.64 EC 5.2.1.7 LCA2p63 mRPE65 RBP-binding membrane protein rd12 Retinal pigment epithelium-specific 65 kDa protein retinal pigment epithelium-specific protein (65kD) retinal pigment epithelium-specific protein 65kDa retinitis pigmentosa 20 (autosomal recessive) retinoid isomerohydrolase Retinol isomerase RP20 sRPE65; BCO family, member 3; Lutein isomerase; Meso-zeaxanthin isomerase; p63; RBP-binding membrane protein; retinal pigment epithelium specific protein 65; Retinal pigment epithelium-specific 65 kDa protein; retinal pigment epithelium-specific protein 65kDa; retinitis pigmentosa 20 (autosomal recessive); Retinoid isomerohydrolase; Retinol isomerase
Gene Aliases: BCO3; LCA2; mRPE65; rd12; RP20; RPE65; sRPE65
UniProt ID: (Human) Q16518
Entrez Gene ID: (Human) 6121
Molecular Function: oxidoreductase oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.