Invitrogen
Manufacturer Code:MA116578
Catalog # AMPA6578
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 401.8B11.3D9 |
Immunogen | Bovine RPE65 microsomal membrane proteins. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C or -80°C if preferred |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: All-trans-retinyl-palmitate hydrolase; BCO family, member 3; LCA2 Leber congenital amaurosis Retinal pigment epithelium-specific 65 kDa protein Retinal pigment epithelium-specific protein Retinitis pigmentos; Lutein isomerase; Meso-zeaxanthin isomerase; modifier of retinal degeneration 1; p63; RBP-binding membrane protein; retinal pigment epithelium 65-like; retinal pigment epithelium abundant protein RPE65; retinal pigment epithelium specific protein 65; retinal pigment epithelium, 65 kDa; Retinal pigment epithelium-specific 65 kDa protein; retinal pigment epithelium-specific 65kD protein; retinal pigment epithelium-specific protein (65kD); retinal pigment epithelium-specific protein 65kDa; retinitis pigmentosa 20 (autosomal recessive); Retinoid isomerohydrolase; Retinol isomerase; RPE65; rpe65 gene for retinal pigment epithelium-specific protein; RPE65, retinoid isomerohydrolase; truncated RPE65
Gene Aliases: 65kDa; A930029L06Rik; BCO3; LCA2; Mord1; mRPE65; rd12; RP20; RPE65; sRPE65
UniProt ID: (Chicken) Q9YGX2, (Bovine) Q28175, (Dog) Q9TVB8, (Human) Q16518, (Mouse) Q91ZQ5, (Rat) O70276
Entrez Gene ID: (Chicken) 395700, (Pig) 100516743, (Bovine) 282043, (Dog) 403803, (Human) 6121, (Mouse) 19892, (Rat) 89826
Molecular Function: oxidoreductase oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.