Invitrogen
Manufacturer Code:PA518890
Catalog # PIPA518890
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (EVKKNARKAPQPE) corresponding to the internal amino acids of RPE65 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: All-trans-retinyl-palmitate hydrolase; Lutein isomerase; Meso-zeaxanthin isomerase; mRPE65 sRPE65 retinal pigment epithelium-specific protein 65kDa LCA2 RP20 rd12 RBP-binding membrane protein p63 retinal pigment epithelium-specific 61 kDa protein retinal pigment epithelium-specific protein (65kD) retinitis pigmentosa 20; retinal pigment epithelium 65-like; Retinal pigment epithelium-specific 65 kDa protein; Retinoid isomerohydrolase; Retinol isomerase
Gene Aliases: Rpe65
UniProt ID: (Rat) O70276
Entrez Gene ID: (Rat) 89826
Molecular Function: oxidoreductase oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.