Invitrogen
Manufacturer Code:PA5110315
Catalog # PIPA5110315
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 274-533 of human RPE65 (NP_000320.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 65kDa A930029L06Rik All-trans-retinyl-palmitate hydrolase BCO3 BOS_3433 LCA2 Lutein isomerase Meso-zeaxanthin isomerase Mord1 mRPE65 rd12 Retinal pigment epithelium-specific 65 kDa protein Retinitis pigmentos Retinoid isomerohydrolase Retinol isomerase RP20 RPE65 sRPE65; All-trans-retinyl-palmitate hydrolase; BCO family, member 3; Lutein isomerase; Meso-zeaxanthin isomerase; modifier of retinal degeneration 1; p63; RBP-binding membrane protein; retinal pigment epithelium 65-like; retinal pigment epithelium specific protein 65; retinal pigment epithelium, 65 kDa; Retinal pigment epithelium-specific 65 kDa protein; retinal pigment epithelium-specific protein 65kDa; retinitis pigmentosa 20 (autosomal recessive); Retinoid isomerohydrolase; Retinol isomerase
Gene Aliases: 65kDa; A930029L06Rik; BCO3; LCA2; Mord1; mRPE65; rd12; RP20; RPE65; sRPE65
UniProt ID: (Human) Q16518, (Mouse) Q91ZQ5, (Rat) O70276
Entrez Gene ID: (Human) 6121, (Mouse) 19892, (Rat) 89826
Molecular Function: oxidoreductase oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.