Novus Biologicals
Manufacturer Code:NBP156853
Catalog # NBP156853
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to RPE(ribulose-5-phosphate-3-epimerase) The peptide sequence was selected from the middle region of RPE. Peptide sequence MMVSKPEQWVKPMAVAGANQYTFHLEATENPGALIKDIRENGMKVGLAIK. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 5.1.3.1 MGC2636 Ribulose-5-phosphate-3-epimerase ribulose-phosphate 3-epimerase RPE2-1; Ribulose-5-phosphate-3-epimerase; Ribulose-phosphate 3-epimerase
Gene Aliases: HUSSY-17; RPE; RPE2-1
UniProt ID: (Human) Q96AT9
Entrez Gene ID: (Human) 6120
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.