Proteintech
Manufacturer Code:121682AP150UL
Catalog # 50-172-7764
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RPE Fusion Protein Ag2810 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ribulose 5 phosphate 3 epimerase Ribulose phosphate 3 epimerase RPE RPE2 1; Ribulose-5-phosphate-3-epimerase; Ribulose-5-phosphate-epimerase; Ribulose-phosphate 3-epimerase
Gene Aliases: 2810429B02Rik; 5730518J08Rik; HUSSY-17; RPE; RPE2-1
UniProt ID: (Human) Q96AT9, (Mouse) Q8VEE0
Entrez Gene ID: (Human) 6120, (Mouse) 66646, (Rat) 501157
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.