Novus Biologicals
Manufacturer Code:NBP273942H
Catalog # NBP273942H
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2H1 |
Immunogen | Full length recombinant protein of human RPC62 (NP_006459) produced in HEK293T cells. |
Conjugate | HRP |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: DNA-directed III 62 kDa polypeptide; DNA-directed III 62 kDa polypeptide DNA-directed RNA polymerase III subunit C DNA-directed RNA polymerase III subunit RPC3 polymerase (RNA) III (DNA directed) polypeptide C (62kD) RNA polymerase III 62 kDa subunit RNA polymerase III subunit C3 RPC62RPC3; DNA-directed RNA polymerase III subunit C; DNA-directed RNA polymerase III subunit RPC3; polymerase (RNA) III (DNA directed) polypeptide C (62kD); polymerase (RNA) III subunit C; RNA polymerase III 62 kDa subunit; RNA polymerase III subunit C3; RPC62
Gene Aliases: POLR3C; RPC3; RPC62
UniProt ID: (Human) Q9BUI4
Entrez Gene ID: (Human) 10623
Molecular Function: DNA-directed RNA polymerase RNA binding protein nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.