Novus Biologicals
Manufacturer Code:NBP187786
Catalog # NBP187786
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:FFRSVFYRSYKEQESKKGFDQEEVFEKPTRETCQGMRHAIYDKLDDDGLIAPGVRVSGDDVIIGKTVTLPENEDELESTNRRYTKRDCSTFLRTSE |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DNA-directed RNA polymerase II 140 kDa polypeptide; DNA-directed RNA polymerase II subunit B; DNA-directed RNA polymerase II subunit RPB2; DNA-directed RNA polymerase II subunit RPB2 EC 2.7.7 EC 2.7.7.6 hRPB140 hsRPB2 POL2RB polymerase (RNA) II (DNA directed) polypeptide B (140kD) polymerase (RNA) II (DNA directed) polypeptide B 140kDa RNA polymerase II subunit 2 RNA polymerase II subunit B2 RNA polymerase II subunit2 RNA polymerase II second largest subunit; polymerase (RNA) II (DNA directed) polypeptide B, 140kDa; polymerase (RNA) II subunit B; RNA polymerase II second largest subunit; RNA polymerase II subunit 2; RNA polymerase II subunit B2
Gene Aliases: hRPB140; POL2RB; POLR2B; RPB2
UniProt ID: (Human) P30876
Entrez Gene ID: (Human) 5431
Molecular Function: DNA-directed RNA polymerase RNA binding protein nucleic acid binding nucleotidyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.