Proteintech
Manufacturer Code:151381AP150UL
Catalog # 50-173-0013
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RPAP1 Fusion Protein Ag7286 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KIAA1403 RPAP1; RNA polymerase II-associated protein 1
Gene Aliases: 1190005L06Rik; A730023M06Rik; AU041788; AW107702; KIAA1403; mKIAA1403; RPAP1
UniProt ID: (Human) Q9BWH6, (Mouse) Q80TE0, (Rat) Q3T1I9
Entrez Gene ID: (Human) 26015, (Mouse) 68925, (Rat) 311338
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.