Invitrogen
Manufacturer Code:PA575738
Catalog # PIPA575738
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues in Human RPA4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Replication factor A protein 4; Replication protein A 30 kDa subunit; Replication protein A 30 kDa subunit RP-A p30 Replication factor A protein 4 RF-A protein 4 RPA4; replication protein A complex 34 kd subunit homolog Rpa4; replication protein A4, 30kDa; replication protein A4, 34kDa; RF-A protein 4; RP-A p30
Gene Aliases: HSU24186; RPA4
UniProt ID: (Human) Q13156
Entrez Gene ID: (Human) 29935
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.