OriGene
Manufacturer Code:TA500763
Catalog # 50-167-1905
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI9A1 |
Immunogen | Full length human recombit protein of human RPA2 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: REPA2 RP-A p32 RP-A p34 RPA32; Replication factor A protein 2; Replication protein A 32 kDa subunit; Replication protein A 34 kDa subunit; replication protein A2, 32kDa; RF-A protein 2; RP-A p32; RP-A p34
Gene Aliases: REPA2; RP-A p32; RP-A p34; RPA2; RPA32; RPA34
UniProt ID: (Human) P15927
Entrez Gene ID: (Human) 6118
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.