Abnova
Manufacturer Code:PAB23915
Catalog # 89-125-622
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human RP1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant RP1.
For Research Use Only
Protein Aliases: DCDC4A; ORP1; Oxygen-regulated protein 1; Retinitis pigmentosa 1 protein; Retinitis pigmentosa RP1 protein
Gene Aliases: DCDC4A; ORP1; RP1
UniProt ID: (Human) P56715
Entrez Gene ID: (Human) 6101
Molecular Function:
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.