Invitrogen
Manufacturer Code:MA537757
Catalog # PIMA537757
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1843CT776.78.21 |
Immunogen | recombinant protein of human ROS1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: c-ros c-ros oncogene 1 receptor tyrosine kinase c-Ros receptor tyrosine kinase c-ros-1 heart - derived c - ros - 1 proto - oncogene MCF3 Proto-oncogene c-Ros proto-oncogene c-Ros-1 Proto-oncogene tyrosine-protein kinase ROS receptor tyrosine kinase c-ros oncogene 1 ROS ROS proto-oncogene 1 receptor tyrosine kinase ROS proto-oncogene 1 receptor tyrosine kinase Ros1 Ros-1 Ros1 proto-oncogene ROS1C RP1-179P9.1 transmembrane tyrosine-specific protein kinase v-ros avian UR2 sarcoma virus oncogene homolog 1 v-ros UR2 sarcoma virus oncogene homolog 1; c-ros oncogene 1 , receptor tyrosine kinase; c-Ros receptor tyrosine kinase; Proto-oncogene c-Ros; Proto-oncogene c-Ros-1; Proto-oncogene tyrosine-protein kinase ROS; Receptor tyrosine kinase c-ros oncogene 1; ROS proto-oncogene 1 , receptor tyrosine kinase; transmembrane tyrosine-specific protein kinase; v-ros avian UR2 sarcoma virus oncogene homolog 1
Gene Aliases: c-ros-1; MCF3; ROS; ROS1
UniProt ID: (Human) P08922
Entrez Gene ID: (Human) 6098
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.