Invitrogen
Manufacturer Code:PA5110313
Catalog # PIPA5110313
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence within amino acids 2100-2200 of human ROS1 (NP_002935.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: c-ros c-Ros receptor tyrosine kinase c-ros-1 MCF3 Proto-oncogene c-Ros Proto-oncogene c-Ros-1 Proto-oncogene tyrosine-protein kinase ROS Receptor tyrosine kinase c-ros oncogene 1 ROS ROS1 Ros-1 ROS1C RP1-179P9.1; c-ros oncogene 1 , receptor tyrosine kinase; c-Ros receptor tyrosine kinase; c-ros-1; Proto-oncogene c-Ros; Proto-oncogene c-Ros-1; Proto-oncogene tyrosine-protein kinase ROS; Receptor tyrosine kinase c-ros oncogene 1; ROS proto-oncogene 1 , receptor tyrosine kinase; transmembrane tyrosine-specific protein kinase; v-ros avian UR2 sarcoma virus oncogene homolog 1
Gene Aliases: c-ros; c-ros-1; MCF3; ROS; Ros-1; ROS1
UniProt ID: (Human) P08922, (Mouse) Q78DX7
Entrez Gene ID: (Human) 6098, (Mouse) 19886
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.