Invitrogen
Manufacturer Code:PA5106255
Catalog # PIPA5106255
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human ROS corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: c-ros c-ros oncogene 1 receptor tyrosine kinase c-Ros receptor tyrosine kinase c-ros-1 heart - derived c - ros - 1 proto - oncogene MCF3 Proto-oncogene c-Ros proto-oncogene c-Ros-1 Proto-oncogene tyrosine-protein kinase ROS receptor tyrosine kinase c-ros oncogene 1 ROS ROS proto-oncogene 1 receptor tyrosine kinase ROS proto-oncogene 1 receptor tyrosine kinase Ros1 Ros-1 Ros1 proto-oncogene ROS1C RP1-179P9.1 transmembrane tyrosine-specific protein kinase v-ros avian UR2 sarcoma virus oncogene homolog 1 v-ros UR2 sarcoma virus oncogene homolog 1; c-ros oncogene 1 , receptor tyrosine kinase; c-Ros receptor tyrosine kinase; c-ros-1; heart - derived c - ros - 1 proto - oncogene; Proto-oncogene c-Ros; Proto-oncogene c-Ros-1; Proto-oncogene tyrosine-protein kinase ROS; Receptor tyrosine kinase c-ros oncogene 1; ROS proto-oncogene 1 , receptor tyrosine kinase; Ros1 proto-oncogene; transmembrane tyrosine-specific protein kinase; v-ros avian UR2 sarcoma virus oncogene homolog 1; v-ros UR2 sarcoma virus oncogene homolog 1
Gene Aliases: c-ros-1; MCF3; ROS; ROS1; ROS1C
UniProt ID: (Human) P08922, (Rat) Q63132
Entrez Gene ID: (Human) 6098, (Rat) 25346
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.