Invitrogen
Manufacturer Code:PA585069
Catalog # PIPA585069
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human ROR alpha. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp686M2414 antibody MGC119326 antibody MGC119329 antibody NR1F1 antibody ROR1 antibody ROR2 antibody ROR3 antibody RZR-ALPHA antibody RZRA antibody RORA antibody nuclear receptor subfamily 1 group F member 1 antibody retinoic acid receptor-related orphan receptor alpha antibody transcription factor RZR-alpha antibody nuclear receptor RZR-alpha antibody retinoid-related orphan receptor alpha antibody ROR-alpha antibody thyroid hormone nuclear receptor alpha variant 4 antibody nuclear receptor ROR-alpha antibody RAR-related orphan receptor A antibody; Nuclear receptor ROR-alpha; Nuclear receptor RZR-alpha; Nuclear receptor subfamily 1 group F member 1; RAR-related orphan receptor A; retinoic acid receptor-related orphan receptor alpha; retinoid-related orphan receptor alpha; Retinoid-related orphan receptor-alpha; ROR-alpha; thyroid hormone nuclear receptor alpha variant 4; transcription factor RZR-alpha
Gene Aliases: 9530021D13Rik; nmf267; NR1F1; ROR1; ROR2; ROR3; RORA; RZR-ALPHA; RZRA; sg; staggerer; tmgc26
UniProt ID: (Human) P35398, (Mouse) P51448
Entrez Gene ID: (Human) 6095, (Mouse) 19883
Molecular Function: nuclear hormone receptor nucleic acid binding receptor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.