Abnova
Manufacturer Code:PAB24915
Catalog # 89-126-589
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to RORA. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of RORA.
For Research Use Only
Protein Aliases: DKFZp686M2414; MGC119326; MGC119329; NR1F1; Nuclear receptor ROR-alpha; Nuclear receptor RZR-alpha; Nuclear receptor subfamily 1 group F member 1; RAR-related orphan receptor A; retinoic acid receptor-related orphan receptor alpha; retinoid-related orphan receptor alpha; Retinoid-related orphan receptor-alpha; ROR-alpha; ROR1; ROR2; ROR3; RZR-ALPHA; RZRA; thyroid hormone nuclear receptor alpha variant 4; transcription factor RZR-alpha
Gene Aliases: NR1F1; ROR1; ROR2; ROR3; RORA; RZR-ALPHA; RZRA
UniProt ID: (Human) P35398
Entrez Gene ID: (Human) 6095
Molecular Function: nuclear hormone receptor nucleic acid binding receptor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.