R&D Systems
Manufacturer Code:MAB20641SP
Catalog # MAB20641SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 231509 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse ROR2 Val28-Gly403 Accession # Q01974 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse MonoclonalAntibody
For Research Use Only
Protein Aliases: BDB BDB1 EC 2.7.10.1 MGC163394 neurotrophic tyrosine kinase receptor-related 2 Neurotrophic tyrosine kinase receptor-related 2 NTRKR2 NTRKR2tyrosine-protein kinase transmembrane receptor ROR2 receptor tyrosine kinase-like orphan receptor 2; neurotrophic tyrosine kinase receptor-related 2; Neurotrophic tyrosine kinase, receptor-related 2; receptor tyrosine kinase-like orphan receptor 2; Tyrosine-protein kinase transmembrane receptor ROR2
Gene Aliases: BDB; BDB1; NTRKR2; ROR2
UniProt ID: (Human) Q01974
Entrez Gene ID: (Human) 4920
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.