R&D Systems
Manufacturer Code:IC9125A025
Catalog # IC9125A025
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 1181A |
Immunogen | <em>E.coli</em> derived recombinant human ROR gamma T/RORC2/NR1F3 Met1-Gln100 Accession # P51449 |
Conjugate | APC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8 degreesC as supplied. |
Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: Nuclear receptor ROR-gamma; Nuclear receptor RZR-gamma; Nuclear receptor subfamily 1 group F member 3; RAR-related orphan nuclear receptor variant 2; RAR-related orphan receptor C; RAR-related orphan receptor C, isoform a; retinoic acid-binding receptor gamma; retinoid-related orphan receptor gamma; Retinoid-related orphan receptor-gamma
Gene Aliases: IMD42; NR1F3; RORC; RORG; RZR-GAMMA; RZRG; TOR
UniProt ID: (Human) P51449
Entrez Gene ID: (Human) 6097
Molecular Function: nuclear hormone receptor nucleic acid binding receptor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.