Invitrogen
Manufacturer Code:17698180
Catalog # 50-112-8643
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | B2D |
Conjugate | APC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: MGC129539 NR1F3 RORG RZR-GAMMA RZRG TOR RORgt; Nuclear receptor ROR-gamma; Nuclear receptor RZR-gamma; Nuclear receptor subfamily 1 group F member 3; RAR-related orphan receptor C; Retinoid-related orphan receptor-gamma; Thymus orphan receptor; TOR
Gene Aliases: Nr1f3; Rorc; Rorg; RORgamma; Thor; TOR
UniProt ID: (Mouse) P51450
Entrez Gene ID: (Mouse) 19885
Molecular Function: nuclear hormone receptor nucleic acid binding receptor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.