Invitrogen
Manufacturer Code:14698837
Catalog # 14-698-837
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | AFKJS-9 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: EGK_01272 IMD42 MGC129539 NR1F3 Nuclear receptor ROR-gamma nuclear receptor RZR-gamma Nuclear receptor subfamily 1 group F member 3 RAR related orphan receptor C RAR-related orphan nuclear receptor variant 2 RAR-related orphan receptor C RAR-related orphan receptor C isoform a RAR-related orphan receptor gamma retinoic acid-binding receptor gamma retinoid-related orphan receptor gamma retinoid-related orphan receptor-gamma ROR Gamma Rorc RORG RORgamma RP11-98D18.11-001 RZRG RZR-GAMMA Thor Thymus orphan receptor TOR; Nuclear receptor ROR-gamma; Nuclear receptor RZR-gamma; Nuclear receptor subfamily 1 group F member 3; RAR-related orphan nuclear receptor variant 2; RAR-related orphan receptor C; RAR-related orphan receptor C, isoform a; retinoic acid-binding receptor gamma; retinoid-related orphan receptor gamma; Retinoid-related orphan receptor-gamma; Thymus orphan receptor; TOR
Gene Aliases: EGK_01272; IMD42; NR1F3; RORC; RORG; RORgamma; RZR-GAMMA; RZRG; Thor; TOR
UniProt ID: (Human) P51449, (Mouse) P51450
Entrez Gene ID: (Rhesus monkey) 717052, (Human) 6097, (Mouse) 19885
Molecular Function: nuclear hormone receptor nucleic acid binding receptor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.