Novus Biologicals
Manufacturer Code:NBP224449
Catalog # NBP224449
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4G419 |
Immunogen | A portion of amino acids 1-50 of human ROR gamma was used as the immunogen. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: MGC129539 mouse ror gamma Nuclear receptor ROR gamma Nuclear receptor ROR-gamma Nuclear receptor RZR gamma Nuclear receptor RZR-gamma Nuclear receptor subfamily 1 group F member 3 RAR-Related Orphan Nuclear Receptor Variant 2 RAR-related orphan receptor C RAR-related orphan receptor gamma retinoic acid-binding receptor gamma retinoid-related orphan receptor gamma Retinoid-related orphan receptor-gamma ROR gamma RORG RORGMGC129539 RZRG RZRGRZR-GAMMA TOR; Nuclear receptor ROR-gamma; Nuclear receptor RZR-gamma; Nuclear receptor subfamily 1 group F member 3; RAR-related orphan nuclear receptor variant 2; RAR-related orphan receptor C; RAR-related orphan receptor C, isoform a; retinoic acid-binding receptor gamma; retinoid-related orphan receptor gamma; Retinoid-related orphan receptor-gamma
Gene Aliases: IMD42; NR1F3; RORC; RORG; RZR-GAMMA; RZRG; TOR
UniProt ID: (Human) P51449
Entrez Gene ID: (Human) 6097
Molecular Function: nuclear hormone receptor nucleic acid binding receptor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.