Invitrogen
Manufacturer Code:PA523148
Catalog # PIPA523148
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide (2 DRAPQRQHRASRELLAAKKTHTSQIE 27) of Retinoid-Related Orphan Receptor Gamma (RORg/NR1F3) protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Nuclear receptor ROR-gamma; Nuclear receptor RZR-gamma; Nuclear receptor subfamily 1 group F member 3; RAR-related orphan nuclear receptor variant 2; RAR-related orphan receptor C; RAR-related orphan receptor C, isoform a; retinoic acid-binding receptor gamma; retinoid-related orphan receptor gamma; Retinoid-related orphan receptor-gamma; RP11-98D18.11-001 NR1F3 RORG RZR-GAMMA RZRG TOR RAR-related orphan nuclear receptor variant 2 nuclear receptor ROR-gamma nuclear receptor RZR-gamma nuclear receptor subfamily 1 group F member 3 retinoic acid-binding receptor gamma retinoid-related orphan receptor gamma; Thymus orphan receptor; TOR
Gene Aliases: IMD42; NR1F3; RORC; RORG; RORgamma; RZR-GAMMA; RZRG; Thor; TOR
UniProt ID: (Human) P51449, (Mouse) P51450
Entrez Gene ID: (Human) 6097, (Mouse) 19885
Molecular Function: nuclear hormone receptor nucleic acid binding receptor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.