Proteintech
Manufacturer Code:110531AP150UL
Catalog # 50-172-6833
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RON MST1R Fusion Protein Ag1537 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: c-met-related tyrosine kinase; CD136; CD136 CDw136 MSP receptor MST1R p185 Ron Protein tyrosine kinase 8 PTK8 RON; CDw136; friend virus susceptibility 2; Macrophage-stimulating protein receptor; Macrophage-stimulating protein receptor alpha chain; Macrophage-stimulating protein receptor beta chain; MSP receptor; p185-Ron; protein tyrosine kinase 8; Protein-tyrosine kinase 8; PTK8 protein tyrosine kinase 8; receptor protein tyrosine kinase, c-met-related; Stem cell-derived tyrosine kinase
Gene Aliases: CD136; CDw136; Fv-2; Fv2; MST1R; PTK8; RON; STK
UniProt ID: (Human) Q04912, (Mouse) E9QMZ4
Entrez Gene ID: (Human) 4486, (Rat) 300999, (Mouse) 19882
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.