Invitrogen
Manufacturer Code:MA531073
Catalog # PIMA531073
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4 |
Immunogen | Recombit Human CD136/MST1R protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: c-met-related tyrosine kinase; CD136; CD136 CDw136 PTK8 RON MSP receptor MST1R variant RON30 MST1R variant RON62 PTK8 protein tyrosine kinase 8 RON variant E2E3 c-met-related tyrosine kinase macrophage-stimulating protein receptor p185-Ron soluble RON variant 1 soluble RON variant 2 soluble RON variant 3 soluble R; CDw136; Macrophage-stimulating protein receptor; Macrophage-stimulating protein receptor alpha chain; Macrophage-stimulating protein receptor beta chain; MSP receptor; p185-Ron; Protein-tyrosine kinase 8; PTK8 protein tyrosine kinase 8
Gene Aliases: CD136; CDw136; MST1R; PTK8; RON
UniProt ID: (Human) Q04912
Entrez Gene ID: (Human) 4486
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.