Invitrogen
Manufacturer Code:PA571878
Catalog # PIPA571878
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 1013-1041 amino acids from the C-terminal region of mouse Mst1r |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD136; CD136 CDw136 PTK8 RON MSP receptor MST1R variant RON30 MST1R variant RON62 PTK8 protein tyrosine kinase 8 RON variant E2E3 c-met-related tyrosine kinase macrophage-stimulating protein receptor p185-Ron soluble RON variant 1 soluble RON variant 2 soluble RON variant 3 soluble R; friend virus susceptibility 2; Macrophage-stimulating protein receptor; Macrophage-stimulating protein receptor alpha chain; Macrophage-stimulating protein receptor beta chain; MSP receptor; p185-Ron; receptor protein tyrosine kinase, c-met-related; Stem cell-derived tyrosine kinase
Gene Aliases: CD136; CDw136; Fv-2; Fv2; Mst1r; PTK8; Ron; STK
UniProt ID: (Mouse) E9QMZ4
Entrez Gene ID: (Mouse) 19882
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.