Novus Biologicals
Manufacturer Code:NBP273928B
Catalog # NBP273928B
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2C12 |
Immunogen | Full length recombinant protein of human ROMO1 (NP_542786) produced in HEK293T cells. |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: Epididymis tissue protein Li 175; Glyrichin; Mitochondrial targeting GxxxG motif protein; mitochondrial targeting GXXXG protein; MTGM; PCM19; Protein MGR2 homolog; Reactive oxygen species modulator 1; ROS modulator 1
Gene Aliases: bA353C18.2; C20orf52; MTGM; MTGMP; ROMO1
UniProt ID: (Human) P60602
Entrez Gene ID: (Human) 140823
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.