OriGene
Manufacturer Code:CF505583
Catalog # 50-166-3751
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1C7 |
Immunogen | Full length human recombit protein of human ROMO1 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: C20orf52 MTGM MTGMP bA353C18.2; Epididymis tissue protein Li 175; Glyrichin; Mitochondrial targeting GxxxG motif protein; mitochondrial targeting GXXXG protein; MTGM; PCM19; Protein MGR2 homolog; Reactive oxygen species modulator 1; ROS modulator 1
Gene Aliases: bA353C18.2; C20orf52; MTGM; MTGMP; ROMO1
UniProt ID: (Human) P60602
Entrez Gene ID: (Human) 140823
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.