Invitrogen
Manufacturer Code:PA570392
Catalog # PIPA570392
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human ROMO1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Epididymis tissue protein Li 175; Glyrichin; Mitochondrial targeting GxxxG motif protein; mitochondrial targeting GXXXG protein; MTGM; PCM19; Protein MGR2 homolog; Reactive oxygen species modulator 1; ROS modulator 1
Gene Aliases: 2010100O12Rik; AI853864; bA353C18.2; C13H20ORF52; C20orf52; MTGM; MTGMP; RA_r333_jsm669B4f; ROMO1; zgc:73345; zgc:77111
UniProt ID: (Bovine) Q3SZV8, (Human) P60602, (Mouse) P60603, (Zebrafish) Q6NYD1
Entrez Gene ID: (Rabbit) 100328919, (Bovine) 618530, (Dog) 477215, (Human) 140823, (Horse) 100069618, (Guinea pig) 100734339, (Rat) 679572, (Mouse) 67067, (Zebrafish) 393770
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.