Novus Biologicals
Manufacturer Code:NB100592
Catalog # NB100592
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residues 1 and 100 of human Rho-associated coiled-coil containing protein kinase 2 using the number given in entry NP_004841.1 (GeneID 9475). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: coiled-coil-containing protein kinase 2 EC 2.7.11 EC 2.7.11.1 KIAA0619 p164 ROCK-2 Rho kinase 2 rho-associated protein kinase 2 Rho-associated coiled-coil containing protein kinase 2; p164 ROCK-2; Rho kinase 2; Rho-associated protein kinase 2; Rho-associated, coiled-coil-containing protein kinase 2; Rho-associated, coiled-coil-containing protein kinase II; ROCK-II
Gene Aliases: KIAA0619; ROCK-II; ROCK2
UniProt ID: (Human) O75116
Entrez Gene ID: (Human) 9475
Molecular Function: kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.