Invitrogen
Manufacturer Code:MA527779
Catalog # PIMA527779
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | GT464 |
Immunogen | Recombit protein encompassing a sequence within the N-terminus region of human ROCK1. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Liver regeneration-related protein LRRG199; p150 RhoA-binding kinase ROK beta; p160 ROCK-1; p160ROCK; P160ROCK Rho-associated coiled-coil containing protein kinase 1 ROCK-I; Renal carcinoma antigen NY-REN-35; Rho-associated coiled-coil forming kinase 1; Rho-associated kinase beta; Rho-associated protein kinase 1; Rho-associated, coiled-coil containing protein kinase 1; Rho-associated, coiled-coil-containing protein kinase 1; Rho-associated, coiled-coil-containing protein kinase I; ROCK-I
Gene Aliases: 1110055K06Rik; Ac2-154; P160ROCK; ROCK-I; ROCK1
UniProt ID: (Human) Q13464, (Mouse) P70335, (Rat) Q63644
Entrez Gene ID: (Human) 6093, (Mouse) 19877, (Rat) 81762
Molecular Function: kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.