Novus Biologicals
Manufacturer Code:NBP242918
Catalog # NBP242918
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 464 |
Immunogen | Recombinant protein encompassing a sequence within the N-terminus region of human ROCK1. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: EC 2.7.11 EC 2.7.11.1 MGC131603 MGC43611 p160 ROCK-1 p160ROCK p160-ROCK PRO0435 Renal carcinoma antigen NY-REN-35 Rho kinase rho-associated protein kinase 1 Rho-associated coiled-coil containing protein kinase 1 Rho-associated coiled-coil-containing protein kinase 1; p160 ROCK-1; p160ROCK; Renal carcinoma antigen NY-REN-35; Rho-associated protein kinase 1; Rho-associated, coiled-coil containing protein kinase 1; Rho-associated, coiled-coil-containing protein kinase 1; Rho-associated, coiled-coil-containing protein kinase I; ROCK-I
Gene Aliases: P160ROCK; ROCK-I; ROCK1
UniProt ID: (Human) Q13464
Entrez Gene ID: (Human) 6093
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.