Invitrogen
Manufacturer Code:PA532219
Catalog # PIPA532219
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to a region within amino acids 1272 and 1366 of Human ROCK1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: p160 ROCK-1; p160ROCK; P160ROCK ROCK-I Rho kinase p160 ROCK-1 p160-ROCK renal carcinoma antigen NY-REN-35 rho-associated protein kinase 1 rho-associated coiled-coil-containing protein kinase 1 rho-associated coiled-coil-containing protein kinase I; Renal carcinoma antigen NY-REN-35; Rho-associated protein kinase 1; Rho-associated, coiled-coil containing protein kinase 1; Rho-associated, coiled-coil-containing protein kinase 1; Rho-associated, coiled-coil-containing protein kinase I; ROCK-I
Gene Aliases: P160ROCK; ROCK-I; ROCK1
UniProt ID: (Human) Q13464
Entrez Gene ID: (Human) 6093
Molecular Function: kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.