Invitrogen
Manufacturer Code:PA542723
Catalog # PIPA542723
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human ROBO2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ast; astray; roundabout 2; Roundabout axon guidance receptor homolog 2 (Drosophila) KIAA1568 SAX3; Roundabout homolog 2; roundabout homolog 2-like; roundabout, axon guidance receptor, homolog 2; zmp:0000000549
Gene Aliases: 2600013A04Rik; 9430089E08Rik; BB097918; D230004I22Rik; KIAA1568; mKIAA1568; ROBO2; SAX3; sb:eu937; unp1881
UniProt ID: (Human) Q9HCK4
Entrez Gene ID: (Human) 6092, (Bovine) 534842, (Rabbit) 100345257, (Dog) 487691, (Horse) 100068957, (Rat) 84409, (Mouse) 268902, (Zebrafish) 60309
Molecular Function:
cell adhesion molecule
cytokine receptor
defense/immunity protein
hydrolase
immunoglobulin receptor superfamily
immunoglobulin superfamily cell adhesion molecule
phosphatase
protein phosphatase
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.