Novus Biologicals
Manufacturer Code:NB10096909
Catalog # NB10096909
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-NQSQRPRPTKKHK corresponding to internal region (near C-Terminus) according to NP_002933.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
For Research Use Only
Protein Aliases: Drosophila) homolog 2 roundabout axon guidance receptor homolog 2 (Drosophila); Roundabout homolog 2; roundabout, axon guidance receptor, homolog 2
Gene Aliases: KIAA1568; ROBO2; SAX3
UniProt ID: (Human) Q9HCK4
Entrez Gene ID: (Human) 6092
Molecular Function: cell adhesion molecule cytokine receptor defense/immunity protein hydrolase immunoglobulin receptor superfamily immunoglobulin superfamily cell adhesion molecule phosphatase protein phosphatase receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.