Invitrogen
Manufacturer Code:PA575814
Catalog # PIPA575814
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues in Human ROBO1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Deleted in U twenty twenty; H-Robo-1; Roundabout homolog 1; Roundabout homolog 1 Deleted in U twenty twenty H-Robo-1 ROBO1 DUTT1; roundabout, axon guidance receptor, homolog 1
Gene Aliases: AW494633; AW742721; DUTT1; Gm310; ROBO1; SAX3
UniProt ID: (Human) Q9Y6N7, (Rat) O55005, (Mouse) O89026
Entrez Gene ID: (Human) 6091, (Rat) 58946, (Mouse) 19876
Molecular Function: cell adhesion molecule cytokine receptor defense/immunity protein hydrolase immunoglobulin receptor superfamily immunoglobulin superfamily cell adhesion molecule phosphatase protein phosphatase receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.