Invitrogen
Manufacturer Code:PA565752
Catalog # PIPA565752
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human ROBO1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Deleted in U twenty twenty; H-Robo-1; ROBO1 DUTT1 roundabout axon guidance receptor homolog 1 (Drosophila) HGNC:10249 FLJ21882 SAX3 axon guidance receptor roundabout 1 MGC131599 MGC133277; Roundabout homolog 1; roundabout, axon guidance receptor, homolog 1
Gene Aliases: DUTT1; ROBO1; SAX3
UniProt ID: (Human) Q9Y6N7
Entrez Gene ID: (Human) 6091
Molecular Function: cell adhesion molecule cytokine receptor defense/immunity protein hydrolase immunoglobulin receptor superfamily immunoglobulin superfamily cell adhesion molecule phosphatase protein phosphatase receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.