Abnova
Manufacturer Code:PAB10401
Catalog # 89-114-833
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to human ROBO1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of ROBO1.
For Research Use Only
Protein Aliases: Deleted in U twenty twenty; DUTT1; FLJ21882; H-Robo-1; MGC131599; MGC133277; Roundabout homolog 1; roundabout, axon guidance receptor, homolog 1; SAX3
Gene Aliases: DUTT1; ROBO1; SAX3
UniProt ID: (Human) Q9Y6N7
Entrez Gene ID: (Human) 6091
Molecular Function: cell adhesion molecule cytokine receptor defense/immunity protein hydrolase immunoglobulin receptor superfamily immunoglobulin superfamily cell adhesion molecule phosphatase protein phosphatase receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.