Proteintech
Manufacturer Code:202191AP150UL
Catalog # 50-173-2774
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Deleted in U twenty twenty; Deleted in U twenty twenty DUTT1 H Robo 1 ROBO1 Roundabout homolog 1 SAX3; H-Robo-1; Roundabout homolog 1; roundabout, axon guidance receptor, homolog 1
Gene Aliases: AW494633; AW742721; DUTT1; Gm310; ROBO1; SAX3
UniProt ID: (Human) Q9Y6N7, (Mouse) O89026
Entrez Gene ID: (Human) 6091, (Mouse) 19876
Molecular Function:
cell adhesion molecule
cytokine receptor
defense/immunity protein
hydrolase
immunoglobulin receptor superfamily
immunoglobulin superfamily cell adhesion molecule
phosphatase
protein phosphatase
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.