Invitrogen
Manufacturer Code:MA531529
Catalog # PIMA531529
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | GT479 |
Immunogen | Recombit protein encompassing a sequence within the center region of human RNaseH2B. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: AGS2; Aicardi-Goutieres syndrome 2 protein; Deleted in lymphocytic leukemia 8; DLEU8 Antibody AGS2 Antibody ribonuclease H2 subunit B Antibody RNASEH2B Antibody; Ribonuclease H2 subunit B; ribonuclease H2, subunit B; Ribonuclease HI subunit B; RNase H2 subunit B
Gene Aliases: AGS2; DLEU8; RNASEH2B
UniProt ID: (Human) Q5TBB1
Entrez Gene ID: (Human) 79621
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.