Proteintech
Manufacturer Code:147641AP150UL
Catalog # 50-172-9703
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RNPEP Fusion Protein Ag6526 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Aminopeptidase B; Aminopeptidase B AP B APB Arginine aminopeptidase Arginyl aminopeptidase RNPEP; AP-B; Arginine aminopeptidase; Arginyl aminopeptidase; arginyl aminopeptidase (aminopeptidase B); Cytosol aminopeptidase IV; RNPEP
Gene Aliases: AP-B; APB; RNPEP
UniProt ID: (Human) Q9H4A4, (Rat) O09175, (Mouse) Q8VCT3
Entrez Gene ID: (Human) 6051, (Rat) 81761, (Mouse) 215615
Molecular Function:
hydrolase
metalloprotease
protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.