Invitrogen
Manufacturer Code:PA554600
Catalog # PIPA554600
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human RNMTL1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 16S rRNA (guanosine(1370)-2'-O)-methyltransferase; 16S rRNA [Gm1370] 2'-O-methyltransferase; putative RNA methyltransferase; RNA methyltransferase like 1; RNA methyltransferase-like protein 1; rRNA methyltransferase 3, mitochondrial
Gene Aliases: HC90; MRM3; RMTL1; RNMTL1
UniProt ID: (Human) Q9HC36
Entrez Gene ID: (Human) 55178
Molecular Function: RNA binding protein RNA methyltransferase methyltransferase nucleic acid binding transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.