Invitrogen
Manufacturer Code:PA541778
Catalog # PIPA541778
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human RNMT |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hcm1p; hCMT1; mRNA (guanine-7-)methyltransferase; mRNA (guanine-N(7)-)-methyltransferase; mRNA cap guanine-N7 methyltransferase; mRNA cap methyltransferase; RG7MT1; RNA (guanine-7-) methyltransferase
Gene Aliases: 2610002P10Rik; AI848273; cm1p; CMT1; CMT1c; hCMT1; hMet; KIAA0398; MET; mKIAA0398; RG7MT1; RNMT
UniProt ID: (Human) O43148, (Mouse) Q9D0L8, (Rat) Q5U2U7
Entrez Gene ID: (Dog) 476200, (Bovine) 511555, (Rabbit) 100340576, (Human) 8731, (Horse) 100050190, (Mouse) 67897, (Rat) 291534, (Guinea pig) 100719755
Molecular Function: RNA binding protein RNA methyltransferase methyltransferase nucleic acid binding transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.