Novus Biologicals
Manufacturer Code:NBP297644V
Catalog # NB102924
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human RNMT fragment. |
Conjugate | DyLight 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp686H1252 EC 2.1.1.56 hcm1p hCMT1 hCMT1c hMet KIAA0398 mRNA (guanine-7-)methyltransferase mRNA (guanine-N(7)-)-methyltransferase mRNA cap guanine-N7 methyltransferase mRNA cap methyltransferase RG7MT1MET RNA (guanine-7-) methyltransferase; hcm1p; hCMT1; mRNA (guanine-7-)methyltransferase; mRNA (guanine-N(7)-)-methyltransferase; mRNA cap guanine-N7 methyltransferase; mRNA cap methyltransferase; RG7MT1; RNA (guanine-7-) methyltransferase
Gene Aliases: cm1p; CMT1; CMT1c; hCMT1; hMet; KIAA0398; MET; RG7MT1; RNMT
UniProt ID: (Human) O43148
Entrez Gene ID: (Human) 8731
Molecular Function: RNA binding protein RNA methyltransferase methyltransferase nucleic acid binding transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.