Invitrogen
Manufacturer Code:MA525276
Catalog # PIMA525276
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3F11 |
Immunogen | Full length human recombit protein of RNH1 produced in HEK293T cell |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Placental ribonuclease inhibitor; Placental RNase inhibitor; RAI; Ribonuclease inhibitor; Ribonuclease inhibitor Placental ribonuclease inhibitor Placental RNase inhibitor Ribonuclease/angiogenin inhibitor 1 RAI RNH1 PRI RNH; Ribonuclease/angiogenin inhibitor 1; testicular tissue protein Li 164
Gene Aliases: EGK_05816; PRI; RAI; RNH; RNH1
UniProt ID: (Human) P13489, (Rat) P29315
Entrez Gene ID: (Rhesus monkey) 720791, (Human) 6050, (Dog) 483402, (Rat) 100360501
Molecular Function: nucleic acid binding transcription cofactor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.