Proteintech
Manufacturer Code:660281IG150UL
Catalog # 50-173-6469
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4A1C4 |
Immunogen | RNH1 Fusion Protein Ag18093 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Placental ribonuclease inhibitor; Placental RNase inhibitor; Placental RNase inhibitor PRI RAI Ribonuclease inhibitor RNH RNH1; RAI; Ribonuclease inhibitor; Ribonuclease/angiogenin inhibitor 1; testicular tissue protein Li 164
Gene Aliases: PRI; RAI; RNH; RNH1
UniProt ID: (Human) P13489
Entrez Gene ID: (Human) 6050
Molecular Function: nucleic acid binding transcription cofactor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.