Proteintech
Manufacturer Code:103451AP150UL
Catalog # 50-172-6231
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RNH1 Fusion Protein Ag0350 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Placental ribonuclease inhibitor; Placental RNase inhibitor; Placental RNase inhibitor PRI RAI Ribonuclease inhibitor RNH RNH1; RAI; Ribonuclease inhibitor; Ribonuclease/angiogenin inhibitor 1; testicular tissue protein Li 164
Gene Aliases: AW546468; C80305; PRI; RAI; RNH; RNH1
UniProt ID: (Human) P13489, (Mouse) Q91VI7
Entrez Gene ID: (Human) 6050, (Mouse) 107702
Molecular Function:
nucleic acid binding
transcription cofactor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.